Learn More
Bio Rad Laboratories RESIN CHELEX 100
Supplier:  Bio Rad Laboratories 1422842
Encompass
Media, Ion exchange; Bio-Rad; Chelex 100 resin; Paired iminodiacetate ions coupled to a styrene divinylbenzene support; 500g; Sodium; 200-400 dry mesh; 75-150um wet bead
Catalog No. 50-901-2932