Learn More
Bio Rad Laboratories RESIN CHELEX 100
Catalog No. 509013100
Encompass
Media, Ion exchange; Bio-Rad; Chelex 100 Resin; Contains paired iminodiacetate ions coupled to a styrene divinylbenzene support; Sodium; 500g; 50-100 dry mesh; 300-1180um wet bead
Catalog No. 50-901-3100 Supplier Bio Rad Laboratories Supplier No. 1422822
Media, Ion exchange; Bio-Rad; Chelex 100 Resin; Contains paired iminodiacetate ions coupled to a styrene divinylbenzene support; Sodium; 500g; 50-100 dry mesh; 300-1180um wet bead