Learn More
TOSOH BIOSCIENCE INC Resin Seeker CIEX-96 Well Plate Kit 20uL
Catalog No. NC3728211
Encompass
The Resin Seeker TOYOPEARL CIEX 96-well plate kit is packed with 4 different TOYOPEARL cation exchange resins: TOYOPEARL GigaCap S-650M, TOYOPEARL GigaCap CM-650M, TOYOPEARL SP-550C, and TOYOPEARL CM-650C. All components necessary to run an experiment are included in each kit: a wash plate and collection plate are included in each 96-well plate package.This 96-well plate kit is manufactured by Orochem and sold by Tosoh Bioscience.
Catalog No. NC3728211 Supplier TOSOH BIOSCIENCE INC Supplier No. OC41MDCEX96
The Resin Seeker TOYOPEARL CIEX 96-well plate kit is packed with 4 different TOYOPEARL cation exchange resins: TOYOPEARL GigaCap S-650M, TOYOPEARL GigaCap CM-650M, TOYOPEARL SP-550C, and TOYOPEARL CM-650C. All components necessary to run an experiment are included in each kit: a wash plate and collection plate are included in each 96-well plate package.This 96-well plate kit is manufactured by Orochem and sold by Tosoh Bioscience.