Learn More
Electron Microscopy Sciences Black Bakelite Thermosetting Hot Mount Resin 1 KG
SDP
Catalog No. 501929669
Encompass_Preferred
Black bakelite with carbon filler, thermosetting. SEM examination.
Catalog No. 50-192-9669 Supplier Electron Microscopy Sciences Supplier No. 1216
Black bakelite with carbon filler, thermosetting. SEM examination.