Learn More
Electron Microscopy Sciences Gray Acryl Thermoplastic Hot Mount Resin 1 KG
SDP
Catalog No. 501929666
Encompass_Preferred
Gray acryl with aluminium filler, thermoplastic. For soft materials. Fast mounting also when used as backing.
Catalog No. 50-192-9666 Supplier Electron Microscopy Sciences Supplier No. 1211
Gray acryl with aluminium filler, thermoplastic. For soft materials. Fast mounting also when used as backing.