Learn More
TARGETMOL CHEMICALS INC Resiniferatoxin 1MG
SDP
Catalog No. 502876326
Encompass_Preferred
Resiniferatoxin ((+)-Resiniferatoxin) is a highly potent synthetic TRPV1 agonist that inhibits the production of Th1 cytokines and has anti-inflammatory activity, reducing serum levels of IL-12, INF-gamma, IL-1beta, TNF-alpha, NO, and PGE. Purity 99.12%
Catalog No. 50-287-6326 Supplier TARGETMOL CHEMICALS INC Supplier No. T342951MG
Resiniferatoxin ((+)-Resiniferatoxin) is a highly potent synthetic TRPV1 agonist that inhibits the production of Th1 cytokines and has anti-inflammatory activity, reducing serum levels of IL-12, INF-gamma, IL-1beta, TNF-alpha, NO, and PGE. Purity 99.12%