Learn More
Apexbio Technology LLC Resiniferatoxin 1mg
SDP
Catalog No. 5000087966
Encompass_Preferred
Resiniferatoxin (CAS 57444-62-9) is a potent selective agonist of the transient receptor potential vanilloid 1 (TRPV1) channel derived from plant sources By activating TRPV1 it induces sustained calcium influx in TRPV1-expressing sensory neurons leading to their desensitization or ablation In animal studies resiniferatoxin has been shown to induce long-lasting elimination of TRPV1-positive primary sensory afferents and blunt associated neural reflexes including those mediating cardiac sympathetic input Resiniferatoxin is widely used in research to investigate pain pathways sensory neuron function and neurogenic inflammation
Catalog No. 50-000-87966 Supplier Apexbio Technology LLC Supplier No. BA70121
Resiniferatoxin (CAS 57444-62-9) is a potent selective agonist of the transient receptor potential vanilloid 1 (TRPV1) channel derived from plant sources By activating TRPV1 it induces sustained calcium influx in TRPV1-expressing sensory neurons leading to their desensitization or ablation In animal studies resiniferatoxin has been shown to induce long-lasting elimination of TRPV1-positive primary sensory afferents and blunt associated neural reflexes including those mediating cardiac sympathetic input Resiniferatoxin is widely used in research to investigate pain pathways sensory neuron function and neurogenic inflammation