Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resiniferatoxin | 57444-62-9 | 1 MG
SDP

Catalog No. 5000420256
Encompass_Preferred

Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. It eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period.

  • Purity of 99.77%
  • Molecular weight: 628.71
  • Chemical formula: C37H40O9
  • Appearance: Solid
  • Causes prolonged channel opening and calcium influx
  • Leads to cytotoxicity in TRPV1-positive pain fibers or cell bodies

Catalog No. 50-004-20256 Supplier Medchemexpress LLC Supplier No. HYN23331MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. It eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period.

  • Purity of 99.77%
  • Molecular weight: 628.71
  • Chemical formula: C37H40O9
  • Appearance: Solid
  • Causes prolonged channel opening and calcium influx
  • Leads to cytotoxicity in TRPV1-positive pain fibers or cell bodies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.