Learn More
Medchemexpress LLC Resiniferatoxin | 57444-62-9 | 5 MG
SDP
Catalog No. 5000420373
Encompass_Preferred
Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. This compound eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period.
  • Selective agonist of TRPV1 receptor
  • Isolated from Euphorbia resinifera plant
  • Eliminates TRPV1+ primary sensory afferents
  • Blunts cardiac sympathetic afferent reflex
  • For research use only
Catalog No. 50-004-20373 Supplier Medchemexpress LLC Supplier No. HYN23335MG
Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. This compound eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period.
  • Selective agonist of TRPV1 receptor
  • Isolated from Euphorbia resinifera plant
  • Eliminates TRPV1+ primary sensory afferents
  • Blunts cardiac sympathetic afferent reflex
  • For research use only