Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resiniferatoxin | 57444-62-9 | 99.8% | 628.71 g/mol | C37H40O9 | 10 MG
SDP

Catalog No. 5000289333
Encompass_Preferred

Resiniferatoxin is a highly potent, naturally derived agonist of the transient receptor potential vanilloid 1 (TRPV1) ion channel. It is used in laboratory research to ablate or modulate TRPV1-expressing sensory neurons and to study pain signaling, neurobiology, and cardiac sympathetic afferent reflexes. Supplied as a high-purity solid with storage and solvent guidance, it is intended for research use only (not for human use).

  • Potent TRPV1 agonist for targeted neuronal ablation and modulation.
  • High chemical purity (99.77%) suitable for sensitive assays.
  • Molecular weight 628.71 g/mol and formula C37H40O9.
  • Stable solid; store protected from light at -20°C; in solution, store at -80°C for long-term stability.
  • Soluble in DMSO; in vitro and in vivo vehicle preparation guidance available.
  • Supplied in small milligram-scale quantities appropriate for research applications.

Catalog No. 50-002-89333 Supplier Medchemexpress LLC Supplier No. HYN233310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resiniferatoxin is a highly potent, naturally derived agonist of the transient receptor potential vanilloid 1 (TRPV1) ion channel. It is used in laboratory research to ablate or modulate TRPV1-expressing sensory neurons and to study pain signaling, neurobiology, and cardiac sympathetic afferent reflexes. Supplied as a high-purity solid with storage and solvent guidance, it is intended for research use only (not for human use).

  • Potent TRPV1 agonist for targeted neuronal ablation and modulation.
  • High chemical purity (99.77%) suitable for sensitive assays.
  • Molecular weight 628.71 g/mol and formula C37H40O9.
  • Stable solid; store protected from light at -20°C; in solution, store at -80°C for long-term stability.
  • Soluble in DMSO; in vitro and in vivo vehicle preparation guidance available.
  • Supplied in small milligram-scale quantities appropriate for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.