Learn More
Medchemexpress LLC Resiniferatoxin | 57444-62-9 | MFCD00135927 | 50 MG
SDP
Catalog No. 5000421402
Encompass_Preferred
Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. Resiniferatoxin eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period. It causes extremely prolonged channel opening and calcium influx, leading to cytotoxicity to TRPV1-positive pain fibers or cell bodies.
  • Selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor
  • Isolated from the Euphorbia resinifera plant
  • Eliminates TRPV1+ primary sensory afferents
  • Blunts cardiac sympathetic afferent reflex
  • Causes prolonged channel opening and calcium influx
  • Leads to cytotoxicity to TRPV1-positive pain fibers or cell bodies
Catalog No. 50-004-21402 Supplier Medchemexpress LLC Supplier No. HYN233350MG
Resiniferatoxin is a selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor. It can be isolated from the Euphorbia resinifera plant. Resiniferatoxin eliminates TRPV1+ primary sensory afferents and blunts cardiac sympathetic afferent reflex for a relatively long period. It causes extremely prolonged channel opening and calcium influx, leading to cytotoxicity to TRPV1-positive pain fibers or cell bodies.
  • Selective agonist of the transient receptor potential vanilloid 1 (TRPV1) receptor
  • Isolated from the Euphorbia resinifera plant
  • Eliminates TRPV1+ primary sensory afferents
  • Blunts cardiac sympathetic afferent reflex
  • Causes prolonged channel opening and calcium influx
  • Leads to cytotoxicity to TRPV1-positive pain fibers or cell bodies