Learn More
Medchemexpress LLC Resiquimod | 144875-48-9 | 99.9% | 314.39 | 500 MG
SDP
Catalog No. 5000352249
Encompass_Preferred
Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6, and IFN-α. It is intended for research use only.
  • TLR7/TLR8 agonist
  • Induces upregulation of cytokines like TNF-α, IL-6, and IFN-α
  • Available in solid and solution forms
  • Used in various research areas including anti-infection, immunology/inflammation
  • Can be used to induce systemic lupus erythematosus models
Catalog No. 50-003-52249 Supplier Medchemexpress LLC Supplier No. HY13740500MG
Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6, and IFN-α. It is intended for research use only.
  • TLR7/TLR8 agonist
  • Induces upregulation of cytokines like TNF-α, IL-6, and IFN-α
  • Available in solid and solution forms
  • Used in various research areas including anti-infection, immunology/inflammation
  • Can be used to induce systemic lupus erythematosus models