Learn More
Selleck Chemical LLC Resiquimod 20mg 144875-48-9 R-848
SDP
Catalog No. 501365692
Encompass_Preferred
Resiquimod (R-848) is an immune response modifier that acts as a potent TLR 7/TLR 8 agonist that induces the upregulation of cytokines such as TNF-?, IL-6 and IFN-?. Resiquimod reduces hepatitis C virus (HCV) infection. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5692 Supplier Selleck Chemical LLC Supplier No. S813320MG
Resiquimod (R-848) is an immune response modifier that acts as a potent TLR 7/TLR 8 agonist that induces the upregulation of cytokines such as TNF-?, IL-6 and IFN-?. Resiquimod reduces hepatitis C virus (HCV) infection. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.