Learn More
Medchemexpress LLC HY-13740 100mg , Resiquimod R848;S28463 CAS:144875-48-9 Purity:98%
SDP
Catalog No. 501493020
Encompass_Preferred
Medchemexpress, HY-13740 100mg Resiquimod R848;S28463 CAS:144875-48-9 Formula:C17H22N4O2 Purity:98% Resiquimod is a Toll-like receptor 7 and 8 ( TLR7/TLR8 ) agonist that induces the levels of cytokines such as TNF-α, IL-6 and IFN-α. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3020 Supplier Medchemexpress LLC Supplier No. HY13740100MG
Medchemexpress, HY-13740 100mg Resiquimod R848;S28463 CAS:144875-48-9 Formula:C17H22N4O2 Purity:98% Resiquimod is a Toll-like receptor 7 and 8 ( TLR7/TLR8 ) agonist that induces the levels of cytokines such as TNF-α, IL-6 and IFN-α. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.