Learn More
Medchemexpress LLC HY-13740 25mg , Resiquimod CAS:144875-48-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1374025MG
Encompass_Preferred
HY-13740 25mg Resiquimod CAS: 144875-48-9 Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IFN-α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0095