Learn More
Selleck Chemical LLC Resiquimod
SDP
Catalog No. 5000000286
Encompass_Preferred
Resiquimod (R-848 S28463) is an immune response modifier that acts as a potent TLR 7/TLR 8 agonist that induces the upregulation of cytokines such as TNF- IL-6 and IFN- Resiquimod reduces hepatitis C virus (HCV) infection Phase 2
Catalog No. 50-000-00286 Supplier Selleck Chemical LLC Supplier No. S813310MM/1ML
Resiquimod (R-848 S28463) is an immune response modifier that acts as a potent TLR 7/TLR 8 agonist that induces the upregulation of cytokines such as TNF- IL-6 and IFN- Resiquimod reduces hepatitis C virus (HCV) infection Phase 2