Learn More
TARGETMOL CHEMICALS INC RESIQUIMOD 50MG
SDP
Catalog No. 502576402
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Resiquimod (R848) is an imidazoquinoline amine and Toll-like receptor (TLR) agonist with potential immune response modifying activity. Resiquimod exerts its effect through the TLR signaling pathway by binding to and activating TLR7 and 8 mainly on dendritic cells macrophages and B-lymphocytes. This induces the nuclear translocation of the transcription activator NF-kB as well as activation of other transcription factors. This may lead to an increase in mRNA levels and subsequent production of cytokines especially interferon-alpha (INF-a) and other cytokines thereby enhancing T-helper 1 (Th1) immune responses. In addition topical application of resiquimod appears to activate Langerhans' cells leading to an enhanced activation of T-lymphocytes. purity: 99%
Catalog No. 50-257-6402 Supplier TARGETMOL CHEMICALS INC Supplier No. T696450MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Resiquimod (R848) is an imidazoquinoline amine and Toll-like receptor (TLR) agonist with potential immune response modifying activity. Resiquimod exerts its effect through the TLR signaling pathway by binding to and activating TLR7 and 8 mainly on dendritic cells macrophages and B-lymphocytes. This induces the nuclear translocation of the transcription activator NF-kB as well as activation of other transcription factors. This may lead to an increase in mRNA levels and subsequent production of cytokines especially interferon-alpha (INF-a) and other cytokines thereby enhancing T-helper 1 (Th1) immune responses. In addition topical application of resiquimod appears to activate Langerhans' cells leading to an enhanced activation of T-lymphocytes. purity: 99%