Learn More
Medchemexpress LLC Resiquimod-d5 (R848-d5) | 2252319-44-9 | 99.8% | 319.41 | 1 MG
SDP
Catalog No. 5000351506
Encompass_Preferred
Resiquimod-d5 is deuterium labeled Resiquimod, a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IFN-α. It can be utilized as a tracer and an internal standard in various analytical methods.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist
  • Induces upregulation of cytokines such as TNF-α, IL-6 and IFN-α
Catalog No. 50-003-51506 Supplier Medchemexpress LLC Supplier No. HY13740S1MG
Resiquimod-d5 is deuterium labeled Resiquimod, a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IFN-α. It can be utilized as a tracer and an internal standard in various analytical methods.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist
  • Induces upregulation of cytokines such as TNF-α, IL-6 and IFN-α