Learn More
Cole-Parmer General Resistance DA-74-3X Decade Box0.01%Acc Resistance 7 Decades
Catalog No. 50632291 Shop All Cole-Parmer Products
Encompass_Preferred
RDS SERIES
  • Accuracies 0.1% or 0.025%
  • 4, 5, 6 or 7 decades
  • "Top of the line" GR performance
  • Rotary switches, gold-plated contacts, low contact R
  • Minimum Ohms = 0
  • OEM models available on special order
DA SERIES
  • Accuracies .01% or 0.1%
  • 3, 4,5,6 or 7 decades
  • Less than one-half the cost of RDS units
  • Easy to use, economical thumbwheel switches
  • Two mechanical configurations
    Note: GR Decade Boxes as RTD Simulators When the ultimate in accuracy is required, GR's RTD-100X or RTD-100RTD should be used Thes
  • Catalog No. 50-632-291 Supplier Cole-Parmer Supplier No. 2001898
    RDS SERIES
    • Accuracies 0.1% or 0.025%
    • 4, 5, 6 or 7 decades
    • "Top of the line" GR performance
    • Rotary switches, gold-plated contacts, low contact R
    • Minimum Ohms = 0
    • OEM models available on special order
    DA SERIES
  • Accuracies .01% or 0.1%
  • 3, 4,5,6 or 7 decades
  • Less than one-half the cost of RDS units
  • Easy to use, economical thumbwheel switches
  • Two mechanical configurations
    Note: GR Decade Boxes as RTD Simulators When the ultimate in accuracy is required, GR's RTD-100X or RTD-100RTD should be used Thes
  • Product Title