Learn More
Resistance Substitution Box
SDPSCI_ED
Catalog No. S43817
Click to view available options
For Use With (Application):
Research, development, repair work
Catalog No. For Use With (Application)
S43817 Research, development, repair work
1 options
Catalog No. S43817 Supplier Elenco Electronics Inc Supplier No. RS500
Compact size makes efficient use of valuable desktop space.
Delivers a very broad range of resistance, making it excellent for research, development and repair work.
  • Aluminum housed
  • One-watt 1% tolerance resistors Accurate range from 0 to 11.1 megohms, in one-ohm steps
  • Three binding posts allow easy connections to outside sources
Specifications
Product Type Resistance Substitution Box
For Use With (Application) Research, development, repair work