Learn More
MSC Drip Pan, 6.5 gal., Green, Polyethylene, Impact Resistant, 1 ea
Catalog No. 502868267
Encompass
This impact-resistant drip pan is designed for containing liquids, offering a reliable solution for various industrial and laboratory applications. Constructed from durable polyethylene, it provides excellent resistance to impacts and chemical exposure.
  • Capacity: 6.5 gal.
  • Material: Polyethylene
  • Color: Green
  • Impact resistant
Catalog No. 50-286-8267 Supplier MSC Supplier No. 13329792
This impact-resistant drip pan is designed for containing liquids, offering a reliable solution for various industrial and laboratory applications. Constructed from durable polyethylene, it provides excellent resistance to impacts and chemical exposure.
  • Capacity: 6.5 gal.
  • Material: Polyethylene
  • Color: Green
  • Impact resistant