Learn More
Crystal Chem Inc Resistin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80588
Encompass
The Resistin ELISA is used to quantify levels of human resistin in plasma, serum, urine, and fluid using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 20 - 1000 pg/mL. High sensitivity of 12 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7754