Learn More
AdipoGen Resistin (human) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45A0023YEKKI01
Encompass_Preferred
ELISA Assay. Detects human resistin. Does not cross-react with mouse resistin, rat resistin, human RELM-beta, mouse RELM-alpha, mouse RELM-beta, rat RELM-alpha, human leptin or human adiponectin. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum. Range: 0.125 to 8ng/ml. Sensitivity: 100pg/ml. Resistin is an adipocytokine. In rodents, increased resistin-levels impair insulin action, while genetic ablation or down-regulation of the resistin gene improves insulin sensitivity. In humans most reports identified inflammatory cells and bone marrow-derived cells as the main source of resistin, indicating a role in inflammatory responses. Plasma resistin levels were shown to be associated with C-reactive protein (CRP), inflammatory bowel disease, Crohn’s disease, atherosclerosis and acute pancreatitis. Atherosclerosis and obesity are increasingly viewed as chronic inflammatory conditions. Circulating resistin levels were reported to be increased in obesity and diabetes.
Catalog No. 50-205-3313