Learn More
AdipoGen Resistin human rec
SDP
Supplier:  AdipoGen AG40A0010YC010
Encompass_Preferred
Protein. Human resistin aa 19-108 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >95% SDS-PAGE. The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules RELM RELM-alpha/FIZZ 1 and RELM-beta/FIZZ 2 of FIZZ found in inflammatory zone are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Catalog No. 50-256-3063