Learn More
AdipoGen Resistin mouse pAb
SDP
Supplier:  AdipoGen AG25A0014C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse resistin. Detects a band of ~10kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules RELM RELMalpha/FIZZ 1 and RELMbeta/FIZZ 2 of FIZZ found in inflammatory zone are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Catalog No. 50-256-3297