Learn More
AdipoGen Resistin mouse rec
SDP
Supplier:  AdipoGen AG40A0011C050
Encompass_Preferred
Protein. Mouse resistin aa 21-114 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH 7.2. Purity >90% SDS-PAGE. The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules RELM RELM-alpha/FIZZ 1 and RELM-beta/FIZZ 2 of FIZZ found in inflammatory zone are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Catalog No. 50-256-3061