Learn More
AdipoGen anti-Resistin (mouse), mAb (MRES 06) (100 µg)
SDP
Supplier:  AdipoGen AG20A0004C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse resistin. Detects a band of ~14kDa by Western blot. Does not cross-react with human resistin. Isotype: Rat IgG2akappa. Clone: MRES 06. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELMalpha/FIZZ 1 and RELMbeta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Catalog No. 50-205-2790