Learn More
AdipoGen anti-Resistin (rat), mAb (RRES 07) (100 µg)
SDP
Supplier:  AdipoGen AG20A0015C100
Encompass_Preferred
Monoclonal Antibody. Recognizes rat resistin. Detects a band of ~10kDa by Western blot. Does not cross-react with human and mouse resistin. Isotype: Mouse IgG1kappa. Clone: RRES 07. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. The adipocytokine resistin which belongs to a family of cysteine-rich C-terminal proteins known as resistin-like molecules (RELM; RELMalpha/FIZZ 1 and RELMbeta/FIZZ 2) of FIZZ (found in inflammatory zone) are thought to be involved in inflammatory processes. Previous studies in mice showed that resistin impairs glucose tolerance and insulin action. In addition, resistin also inhibits adipogenesis in murine 3T3-L1 cells. Therefore resistin has also been proposed as an adipocyte-secreted factor thought to link obesity and T2DM.
Catalog No. 50-205-2797