Learn More
Cole-Parmer Myron L CS10-T Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable
Catalog No. 50631789 Shop All Cole-Parmer Products
Encompass
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
Catalog No. 50-631-789 Supplier Cole-Parmer Supplier No. 0597072
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.