Learn More
Vernier™ Vernier Resistor Board
SDPSCI_ED
Catalog No. S16016ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
KidWind Wind Turbines and Solar Panels, Energy Sensor
Catalog No. For Use With (Equipment)
S16016ND KidWind Wind Turbines and Solar Panels, Energy Sensor
1 options
Catalog No. S16016ND Supplier Vernier™ Supplier No. VESRB
Add to Cart
Add to Cart
Includes
Vernier Resistor Board, Four insulated jumper wires fitted with mini-alligator clips
The Vernier Resistor Board has seven 1 W 5% resistors mounted on a circuit board. Test clip holes on either side of the resistors allow for easy connections to clips on wire leads.
Resistor values: 10 Ω, 15 Ω, 20 Ω, 30 Ω, 39 Ω, 51 Ω, and 100 Ω Dimensions: 3.5 in long by 2 in wide
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) KidWind Wind Turbines and Solar Panels, Energy Sensor
For Use With (Application) Educational use only
Includes Four insulated jumper wires fitted with mini-alligator clips