Learn More
Medchemexpress LLC HY-12216 100mg , Resmetirom CAS:920509-32-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12216100MG
Encompass_Preferred
HY-12216 100mg Resmetirom CAS: 920509-32-6 Resmetirom (MGL-3196) is a highly selective thyroid hormone receptor β (THR-β) agonist with an EC50 value of 0.21 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9989