Learn More
Selleck Chemical LLC Resmetirom (MGL-3196) 5mg 920509-32-6 VIA-3196
SDP
Supplier:  Selleck Chemical LLC S66635MG
Encompass_Preferred
Resmetirom (MGL-3196, VIA-3196) is a liver-directed, orally active, selective thyroid hormone receptor ?(THR-?) agonist with an EC50 of 0.21 ?M,shows 28-fold selective for THR-? over THR-? in a functional assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1772005