Learn More
Selleck Chemical LLC Resminostat S2693-10mg
SDP
Catalog No. 501362790
Encompass_Preferred
Resminostat (RAS2410) dose-dependently and selectively inhibits HDAC1/3/6 with IC50 of 42 5 nM/50 1 nM/71 8 nM less potent to HDAC8 with IC50 of 877 nM
Catalog No. 50-136-2790 Supplier Selleck Chemical LLC Supplier No. S269310MG
Resminostat (RAS2410) dose-dependently and selectively inhibits HDAC1/3/6 with IC50 of 42 5 nM/50 1 nM/71 8 nM less potent to HDAC8 with IC50 of 877 nM