Learn More
Medchemexpress LLC HY-14718A 10mg Medchemexpress, Resminostat (hydrochloride) CAS:1187075-34-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14718A10MG
Encompass_Preferred
Medchemexpress, HY-14718A 10mg Resminostat (hydrochloride) CAS:1187075-34-8 Resminostat hydrochloride is a potent inhibitor of HDAC1, HDAC3 and HDAC6, with mean IC50 values of 42.5, 50.1, 71.8 nM, respectively, and shows less potent activities against HDAC8, with an IC50 of 877 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1631