Learn More
TARGETMOL CHEMICALS INC RESMINOSTAT HYDROCHLORID 10MG
SDP
Catalog No. 502579895
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Resminostat hydrochloride (RAS2410 hydrochloride) is an effective inhibitor of HDAC1/HDAC3/HDAC6 (IC50 42.5/50.1/71.8 nM) respectively and shows less potent activities against HDAC8 (IC50 877 nM). purity: 99%
Catalog No. 50-257-9895 Supplier TARGETMOL CHEMICALS INC Supplier No. T437010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Resminostat hydrochloride (RAS2410 hydrochloride) is an effective inhibitor of HDAC1/HDAC3/HDAC6 (IC50 42.5/50.1/71.8 nM) respectively and shows less potent activities against HDAC8 (IC50 877 nM). purity: 99%