Learn More
Medchemexpress LLC Resminostat (hydrochloride) | 1187075-34-8 | 99.67% | 385.87 | 25 MG
SDP
Catalog No. 5000363801
Encompass_Preferred
Resminostat hydrochloride is a potent inhibitor of HDAC1, HDAC3, and HDAC6, with IC50 values in the nanomolar range. It is used for research applications and appears as a white to off-white solid. This compound shows less potent activity against HDAC8.
  • Potent inhibitor of HDAC1, HDAC3, and HDAC6
  • Exhibits less potent activity against HDAC8
  • Induces histone acetylation in myeloma cells
  • Inhibits cell growth and induces apoptosis in myeloma cells
  • Synergistic activity with anti-myeloma agents
  • Reduces viability of HCC cells when co-treated with AZD-2014
Catalog No. 50-003-63801 Supplier Medchemexpress LLC Supplier No. 25MGHY14718A
Resminostat hydrochloride is a potent inhibitor of HDAC1, HDAC3, and HDAC6, with IC50 values in the nanomolar range. It is used for research applications and appears as a white to off-white solid. This compound shows less potent activity against HDAC8.
  • Potent inhibitor of HDAC1, HDAC3, and HDAC6
  • Exhibits less potent activity against HDAC8
  • Induces histone acetylation in myeloma cells
  • Inhibits cell growth and induces apoptosis in myeloma cells
  • Synergistic activity with anti-myeloma agents
  • Reduces viability of HCC cells when co-treated with AZD-2014