Learn More
Medchemexpress LLC Resminostat hydrochloride | 1187075-34-8 | 99.67% | 385.87 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14718A5MG
Encompass_Preferred
Resminostat hydrochloride is a potent inhibitor of HDAC1, HDAC3, and HDAC6, with mean IC50 values of 42.5, 50.1, and 71.8 nM, respectively. It exhibits less potent activity against HDAC8, with an IC50 of 877 nM. This product is for research use only and is not sold to patients.
  • Potent inhibitor of HDAC1, HDAC3, and HDAC6
  • IC50 values of 42.5, 50.1, and 71.8 nM for HDAC1, HDAC3, and HDAC6 respectively
  • Less potent activity against HDAC8
  • For research use only
Catalog No. 50-003-62871