Learn More
Apexbio Technology LLC Resminostat (RAS2410), 10mg. Cas: 864814-88-0 MFCD: MFCD18633257. Potent HDAC inhibitor
SDP
Supplier:  Apexbio Technology LLC A410810
Encompass_Preferred
Resminostat, also known as RAS2410, is a potent inhibitor of histone deacetylase (HDAC) classes I and II (including HDAC1, HDAC3 and HDAC6) with 50% inhibition concentration IC50 values ranging from 43 to 72 nmol/L. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4882