Learn More
Cayman Chemical Resminostat (hydrochloride), Purity: ≥95%, CAS: 1187075-34-8, 10 mg
SDP
Catalog No. 502965297
Encompass_Preferred
An orally bioavailable inhibitor of HDAC1, HDAC3, and HDAC6 (IC50s = 43-72 nM), resulting in hyperacetylation of histone H4 in multiple myeloma cells; abrogates cell growth and strongly induces apoptosis in multiple myeloma cells (IC50s = 2.5-3 µM); dose-dependently inhibits HDAC activity in vivo
Catalog No. 50-296-5297 Supplier Cayman Chemical Supplier No. 1755310 MG
An orally bioavailable inhibitor of HDAC1, HDAC3, and HDAC6 (IC50s = 43-72 nM), resulting in hyperacetylation of histone H4 in multiple myeloma cells; abrogates cell growth and strongly induces apoptosis in multiple myeloma cells (IC50s = 2.5-3 µM); dose-dependently inhibits HDAC activity in vivo