Learn More
Cayman Chemical ResmInostathydrochlorIde 50mg
SDP
Catalog No. 502966101
Encompass_Preferred
An orally bioavailable inhibitor of HDAC1, HDAC3, and HDAC6 (IC50s = 43-72 nM), resulting in hyperacetylation of histone H4 in multiple myeloma cells; abrogates cell growth and strongly induces apoptosis in multiple myeloma cells (IC50s = 2.5-3 µM); dose-dependently inhibits HDAC activity in vivo
Catalog No. 50-296-6101 Supplier Cayman Chemical Supplier No. 1755350 MG
An orally bioavailable inhibitor of HDAC1, HDAC3, and HDAC6 (IC50s = 43-72 nM), resulting in hyperacetylation of histone H4 in multiple myeloma cells; abrogates cell growth and strongly induces apoptosis in multiple myeloma cells (IC50s = 2.5-3 µM); dose-dependently inhibits HDAC activity in vivo