Learn More
Waters Corp Resolution Check Standard, GC/MS, Heptacosafluorotributylamine, Liquid
Catalog No. 50466514 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standard is a routine use mass spectrometry standard for all gas chromatography/mass spectrometry products, designed to perform resolution check testing. It contains Heptacosafluorotributylamine.
  • Designed for instrument performance and mass spectrometry detection.
  • Packaged as one per package.
  • Stores at room temperature.
  • Contains hazardous material which requires special freight handling.
Catalog No. 50-466-514 Supplier Waters Corp Supplier No. 186006862
This standard is a routine use mass spectrometry standard for all gas chromatography/mass spectrometry products, designed to perform resolution check testing. It contains Heptacosafluorotributylamine.
  • Designed for instrument performance and mass spectrometry detection.
  • Packaged as one per package.
  • Stores at room temperature.
  • Contains hazardous material which requires special freight handling.