Learn More
Waters Corp Resolve C18 Radial-Pak Column, 90Å, 10 µm, 8 mm X 100 mm, 1/pk
Catalog No. 50785924 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The non-encapped Resolve C18 column offers significantly different selectivity compared to similar HPLC packings like Nova-Pak and µBondapak. The change in chromatographic behavior is most commonly noticed with polar compounds, which are typically more retained.
Catalog No. 50-785-924 Supplier Waters Corp Supplier No. WAT084720
The non-encapped Resolve C18 column offers significantly different selectivity compared to similar HPLC packings like Nova-Pak and µBondapak. The change in chromatographic behavior is most commonly noticed with polar compounds, which are typically more retained.