Learn More
Waters Corp Resolve C18 Column, 90Å, 5 µm, 3.9 mm X 300 mm, 1/pk
Supplier:  Waters Corp WAT011740
Encompass_Preferred
The non-encapped Resolve C18 column offers significantly different selectivity compared to similar HPLC packings like Nova-Pak and µBondapak. The change in chromatographic behavior is most commonly noticed with polar compounds, which are typically more retained.
Catalog No. 50-785-047