Learn More
Waters Corp Resolve C8 Radial-Pak Column, 90Å, 10 µm, 5 mm X 100 mm, 1/pk
Supplier:  Waters Corp WAT085672
Encompass_Preferred
The non-endcapped Resolve packings are significantly different from Waters’ other packing materials. The change in chromatographic behavior is most commonly noticed with polar compounds which are typically more retained.
Catalog No. 50-785-943