Learn More
Waters Corp Waters Resolve Column, Cyano (CN), 10 μm, 8 mm x 100 mm
Catalog No. 50785923 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The non-endcapped Resolve packings are significantly different from other packing materials. The change in chromatographic behavior is most commonly noticed with polar compounds which are typically more retained.
  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • 90 Å pore size
  • Radial-Pak column format
  • Surface area of 200
  • For HPLC systems
  • USP Classification L10
  • 8 mm inner diameter
  • 100 mm length
  • 3% carbon load
  • 1 unit per package
Catalog No. 50-785-923 Supplier Waters Corp Supplier No. WAT084636
The non-endcapped Resolve packings are significantly different from other packing materials. The change in chromatographic behavior is most commonly noticed with polar compounds which are typically more retained.
  • Cyano (CN) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • 90 Å pore size
  • Radial-Pak column format
  • Surface area of 200
  • For HPLC systems
  • USP Classification L10
  • 8 mm inner diameter
  • 100 mm length
  • 3% carbon load
  • 1 unit per package