Learn More
Waters Corp Waters Column, Resolve Silica, 90Å, 5μm, 3.9 mm X 150 mm
Supplier:  Waters Corp WAT086016
Encompass_Preferred
This silica column features non-endcapped Resolve packings, providing distinct chromatographic behavior, especially for polar compounds that exhibit increased retention. It allows for the chromatography of basic chemicals using mobile phase modifiers to mitigate excessive tailing. This column is suitable for high-resolution applications in liquid chromatography.
  • Designed to deliver excellent quality and desirable results for liquid chromatography.
  • Manufactured in specialized, cutting-edge facilities.
  • Ensures high-caliber lab equipment for trustworthy and consistent analytical findings.
  • Key performance features include column efficiency.
  • Offers peak symmetry.
  • Provides column longevity.
  • Ensures column-to-column repeatability.
Catalog No. 50-785-952