Learn More
Cayman Chemical Resolvin D1, 872993-05-0, 25 g
SDP
Supplier:  Cayman Chemical 1001255425
Encompass_Preferred
Molecular Formula C22H32O5, Purity ≥95%, Formula Weight 376.5, Synonyms: 17(S)-Resolvin D1, RvD1; A potent anti-inflammatory mediator derived physiologically from the sequential oxygenation of DHA by 15- and 5-LO; reduces human leukocyte transendothelial migration (EC50 = ~30 nM) and limits leukocyte infiltration in a mouse model of peritonitis.
Catalog No. NC9019976