Learn More
Cayman Chemical Resolvin D1 AChE Tracer, 100 dtn
SDP
Catalog No. 502942381
Encompass_Preferred
Resolvin D1 AChE Tracer has been tested and formulated to work exclusively with Cayman’s Resolvin D1 ELISA Antiserum (Item No. 500380).
Catalog No. 50-294-2381 Supplier Cayman Chemical Supplier No. 400380100 DTN
Resolvin D1 AChE Tracer has been tested and formulated to work exclusively with Cayman’s Resolvin D1 ELISA Antiserum (Item No. 500380).