Learn More
Cayman Chemical ResolvIn D1-d5 25ug
SDP
Catalog No. 502954816
Encompass_Preferred
An internal standard for the quantification of resolvin D1 by GC- or LC-MS
Catalog No. 50-295-4816 Supplier Cayman Chemical Supplier No. 1118225 UG
An internal standard for the quantification of resolvin D1 by GC- or LC-MS