Learn More
Cayman Chemical ResolvIn D1 methyl ester 100UG
SDP
Catalog No. 502893556
Encompass_Preferred
A methyl ester version of the free acid that may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties
Catalog No. 50-289-3556 Supplier Cayman Chemical Supplier No. 9001815100 UG
A methyl ester version of the free acid that may act as a lipophilic prodrug form that will alter its distribution and pharmacokinetic properties